Learn More
Hach Company Hach PVC Pump Tubing, 0.040 in (1.016 mm) ID, White/White, 12-Pack
Catalog No. NC3351002 Shop All Hach Company Products
Encompass
PVC pump tubing with a 0.040 in (1.016 mm) inner diameter, sold as a pack of 12 white tubes for use in peristaltic and low-pressure liquid transfer pumps. Suitable for laboratory and field instrumentation where consistent tubing ID is required for accurate flow control.
  • 0.040 in (1.016 mm) inner diameter for consistent flow.
  • PVC construction suitable for general-purpose liquid transfer.
  • White/White color for easy visual inspection.
  • Pack of 12 tubes for maintenance and replacement needs.
  • Designed for use with peristaltic and low-pressure pumps.
Catalog No. NC3351002 Supplier Hach Company Supplier No. 53409
PVC pump tubing with a 0.040 in (1.016 mm) inner diameter, sold as a pack of 12 white tubes for use in peristaltic and low-pressure liquid transfer pumps. Suitable for laboratory and field instrumentation where consistent tubing ID is required for accurate flow control.
  • 0.040 in (1.016 mm) inner diameter for consistent flow.
  • PVC construction suitable for general-purpose liquid transfer.
  • White/White color for easy visual inspection.
  • Pack of 12 tubes for maintenance and replacement needs.
  • Designed for use with peristaltic and low-pressure pumps.