Learn More
Hach Company PVC Pump Tubing, 0.081 in (2.057 mm) ID, Purple/Purple, 12/Pack - For Lachat Pumps
Catalog No. NC3350985 Shop All Hach Company Products
Encompass
PVC pump tubing with a 0.081 in (2.057 mm) inner diameter, sold as a 12-pack. Designed for peristaltic pump fluid transfer and listed for use with Lachat-style sampling equipment.
  • Inner diameter 0.081 in (2.057 mm).
  • Color purple/purple.
  • Sold 12 per pack.
  • Made from flexible PVC for reliable fluid transfer.
  • Suitable for pump and sampling applications.
Catalog No. NC3350985 Supplier Hach Company Supplier No. 53415
PVC pump tubing with a 0.081 in (2.057 mm) inner diameter, sold as a 12-pack. Designed for peristaltic pump fluid transfer and listed for use with Lachat-style sampling equipment.
  • Inner diameter 0.081 in (2.057 mm).
  • Color purple/purple.
  • Sold 12 per pack.
  • Made from flexible PVC for reliable fluid transfer.
  • Suitable for pump and sampling applications.