Learn More
Thermo Scientific PUMP TUBING SAMPLE ICP 6/PK
Supplier:  Thermo Scientific 842312051511
Encompass
Pump Sample Tube Aqueous; Standard tygon orange/white pump tubing.; One pack of six tubes.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9608336