Learn More
EZKEM Pump Tubing Yellow/Yellow 0.056 ID 6x36 12pk
SDP
Catalog No. NC1718711
Encompass_Preferred
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000353 Yellow/Yellow 0.056 ID 12/pk
Catalog No. NC1718711 Supplier EZKEM Supplier No. A000353
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000353 Yellow/Yellow 0.056 ID 12/pk