Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, PharmaPure, I/P 73; 25 Ft
Catalog No. NC9090781
Encompass
Masterflex PharmaPure Precision pump tubing has ultra-low spallation and low extractability to protect the purity of samples. This high-performance formulation is nontoxic and nonhemolytic making it ideal for sterile, filtration, and cell media applications. Low extractables and low gas permeability provides a protective barrier to chemicals that damage tubing while maintaining the integrity of your fluid.
This item is not returnable. View return policy
Catalog No. NC9090781 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643573/DEL
Masterflex PharmaPure Precision pump tubing has ultra-low spallation and low extractability to protect the purity of samples. This high-performance formulation is nontoxic and nonhemolytic making it ideal for sterile, filtration, and cell media applications. Low extractables and low gas permeability provides a protective barrier to chemicals that damage tubing while maintaining the integrity of your fluid.