Learn More
Hach Company Pump, Vacuum/Pressure, Portable, 220v, 50 Hz
Supplier:  Hach Company 2824802
Encompass
Quiet, pulse less air delivery and vacuum. Compact dimensions (WxDxH) 15x39x20 cm (6x15.4x8 inches). Complete with vacuum and pressure gauges and regulators, oil filter assembly, hose nipples, outlet filter, switch, cord, and plug.Capacity; 0.6 liters/sec (1.3 cfm) at 0 mm Hg. 124 W (1/6 HP). 1725 RPM. European Plug.
Catalog No. 50-199-6727