Learn More
Vici Metronics PUMPARRAY FOR 6 PUMPS, EQUIPPE
SDP
Catalog No. NC1960933
Encompass
PumpArray for 6 pumps, equipped with USB 2.0 port for serial control, 320W power supply, cooling fan, heater controller, cabling, and tubing
Catalog No. NC1960933 Supplier Vici Metronics Supplier No. PUMPARRAY6
PumpArray for 6 pumps, equipped with USB 2.0 port for serial control, 320W power supply, cooling fan, heater controller, cabling, and tubing