Learn More
Essendant PUMPDRUMSPHN2ADPTR
Supplier:  Essendant BWK00423
Encompass_Preferred
Versatile plastic drum pump fits a variety of sizes works with 15 30 and 55-gallon drums Easy fingertip control The easy-to-use screw top vent controls siphon flow Compatible with most cleaning solutions and corrosive chemicals Use with thin easily flowing chemicals Do not use with solvents Available with 2 IPS adaptor for steel and plastic drums or a 2 buttress adaptor for plastic drums with buttress threads Pump Application Type- Plastic Siphon Drum Pump Style- Push Pump Operating Principle- Displacement Volume per Minute- 5 gpm
Catalog No. 50-000-18613