Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Easy-Load Long-Shaft Pump Head for Precision Tubing, Black PPS Housing, SS Rotor
Catalog No. NC1650842
Encompass
Standard heads are precision molded to accommodate a single size of tubing¿use for common fluid transfer applications requiring accuracy and excellent repeatability. Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors; CRS offers economy, while SS offers better chemical resistance and durability. Use with Masterflex I/P pumps requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives. Pump head is compact and simple in design with same side tubing entry/exit¿perfect for space-limited applications. Standard heads are compact and easy to integrate into your OEM applications. Call for more information on our comprehensive OEM services.
Catalog No. NC1650842 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7760162
Standard heads are precision molded to accommodate a single size of tubing¿use for common fluid transfer applications requiring accuracy and excellent repeatability. Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors; CRS offers economy, while SS offers better chemical resistance and durability. Use with Masterflex I/P pumps requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives. Pump head is compact and simple in design with same side tubing entry/exit¿perfect for space-limited applications. Standard heads are compact and easy to integrate into your OEM applications. Call for more information on our comprehensive OEM services.