Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Pump Head Adapter for NEMA Type 56 C Motor, 650 rpm; 4.8:1 Gear Ratio
Catalog No. NC1311660
Encompass
Connects any I/P pump head to a 56C frame motor. Cast aluminum adapters feature built-in gear box for easy setup. Compatible with all I/P pump heads. Works with any 56C frame motor. 4.8:1 gear ratio.
Catalog No. NC1311660 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7749010
Connects any I/P pump head to a 56C frame motor. Cast aluminum adapters feature built-in gear box for easy setup. Compatible with all I/P pump heads. Works with any 56C frame motor. 4.8:1 gear ratio.