Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Pump Head Adapter for NEMA 56C Frame Motor, 3.7:1 Gear Ratio
Catalog No. NC0551228
Encompass
Use adapters to mount MASTERFLEX L/S pump heads to a 56C frame motor (pump head and motor sold separately).
Catalog No. NC0551228 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7749500
Use adapters to mount MASTERFLEX L/S pump heads to a 56C frame motor (pump head and motor sold separately).