Learn More
BURKLE INC PumpMaster acids/ chemical liquids, FKM, green
Catalog No. 5000025834
Encompass_Preferred
The PumpMaster in green for filling and dosing acids and chemical liquids is equipped with high-quality FKM seals. Ideal for the safe handling of light acids and chemicals.
Catalog No. 50-000-25834 Supplier BURKLE INC Supplier No. 52021000
The PumpMaster in green for filling and dosing acids and chemical liquids is equipped with high-quality FKM seals. Ideal for the safe handling of light acids and chemicals.