Learn More
BURKLE INC PumpMaster non-aggressive liquids, PP/EPDM, red
Catalog No. 5000025737
Encompass_Preferred
The red PumpMaster manual overpressure pump is ideal for pumping aqueous, non-aggressive to slightly alkaline liquids. Equipped with durable EPDM seals, it ensures safe and reliable transfer of these liquids. The PumpMaster range offers an easy solution for filling, transferring, and emptying canisters and drums. It comes with three universal plugs that fit container openings from 1.83 to 2.36 inches (46.5 to 60 mm), allowing quick and secure adaptation without exact thread specifications. Perfect for industrial, chemical processing, food production, and laboratory applications requiring safe, manual liquid handling and transfer.
Catalog No. 50-000-25737 Supplier BURKLE INC Supplier No. 52022000
The red PumpMaster manual overpressure pump is ideal for pumping aqueous, non-aggressive to slightly alkaline liquids. Equipped with durable EPDM seals, it ensures safe and reliable transfer of these liquids. The PumpMaster range offers an easy solution for filling, transferring, and emptying canisters and drums. It comes with three universal plugs that fit container openings from 1.83 to 2.36 inches (46.5 to 60 mm), allowing quick and secure adaptation without exact thread specifications. Perfect for industrial, chemical processing, food production, and laboratory applications requiring safe, manual liquid handling and transfer.