Learn More
World Precision Instrument Corneoscleral Punch, 8cm, 1.5mm Wide
SDP
Catalog No. 50822374
Encompass
This corneoscleral punch is an excellent tool for opening the skull of a small animal without damaging the brain or spinal cord.The lower jaw of the punch is a flat plate which can be inserted into the skull with much less damage to the brain than a rongeur.
Catalog No. 50-822-374 Supplier World Precision Instrument Supplier No. 500143
This corneoscleral punch is an excellent tool for opening the skull of a small animal without damaging the brain or spinal cord.The lower jaw of the punch is a flat plate which can be inserted into the skull with much less damage to the brain than a rongeur.