Learn More
Fisherbrand™ Punched Aluminum Test Tube Rack
Catalog No. 01189159 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
24 Slot
25 Slot
40 Slot
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-189-159 25 Slot
01-189-161 24 Slot
01-189-160 40 Slot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-189-159
Add to Cart
Add to Cart
This Punched Aluminum Test Tube Rack features a Z-shape for holding test tubes. Three configurations available: 24, 25, and 40 slots.
  • Aluminum construction
  • Anti-corrosive
  • Low profile and well balanced
  • Available in three configurations: 24, 25, and 40 slots
Specifications
Certifications/Compliance ISO, GMP
Autoclavable Autoclavable
Color Silver
Holds Test Tubes
Material Stainless Steel
Shape Z-shaped
Stackable Yes, When Empty
Sterility Non-sterile
Size 25 Slot
Product Type Test Tube Rack