Learn More
Waters Corp Puncture Needle Assembly, .059 OD, I-Class, M-Class, UPC2 Compatible
Catalog No. 502392973 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Puncture Needle Assembly is designed for use on I-Class, M-Class, and UPC2 Fixed Loop Sample Managers. Its primary function is to pierce vial caps, allowing the sample needle to enter vials for sample analysis.
  • Designed for use on I-Class, M-Class, and UPC2 Fixed Loop Sample Managers
  • Pierces vial caps to allow sample needle to enter vials
  • .059 outer diameter
  • UPLC system compatible
  • Mechanical type
  • One unit per package
  • As required replacement schedule
Catalog No. 50-239-2973 Supplier Waters Corp Supplier No. 700006067
This replacement Puncture Needle Assembly is designed for use on I-Class, M-Class, and UPC2 Fixed Loop Sample Managers. Its primary function is to pierce vial caps, allowing the sample needle to enter vials for sample analysis.
  • Designed for use on I-Class, M-Class, and UPC2 Fixed Loop Sample Managers
  • Pierces vial caps to allow sample needle to enter vials
  • .059 outer diameter
  • UPLC system compatible
  • Mechanical type
  • One unit per package
  • As required replacement schedule