Learn More
Medchemexpress LLC HY-N0063 20mg , Punicalagin CAS:65995-63-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN006320MG
Encompass_Preferred
HY-N0063 20mg Punicalagin CAS: 65995-63-3 Punicalagin is a polyphenol ingredient isolated from Pomegranate (Punica granatum L.) or the leaves of Terminalia catappa L.. Punicalagin is a reversible and non-competitive 3CLpro inhibitor and inhibits SARS-CoV-2 replication in vitro. Punicalagin is an anti-hepatitis B virus (HBV) agent and has antioxidant, anti-inflammatory, and anticancer effects. Punicalagin has the potential for the research of COVID-19. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9406