Learn More
TARGETMOL CHEMICALS INC PUNICALIN 5MG
SDP
Catalog No. 502576717
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Punicalin exerts anti-inflammatory antioxidative and anti-hepatotoxic activities it shows inhibitory activity on HIV-1 reverse transcriptase in a dose-dependent manner with an IC50 of 0.11 microg/ml (0.14 microM). purity: 99%
Catalog No. 50-257-6717 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S17185MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Punicalin exerts anti-inflammatory antioxidative and anti-hepatotoxic activities it shows inhibitory activity on HIV-1 reverse transcriptase in a dose-dependent manner with an IC50 of 0.11 microg/ml (0.14 microM). purity: 99%