Learn More
Medchemexpress LLC Punicalin | 65995-64-4 | 99.9% | 782.53 g/mol | C34H22O22 | 10 MG
SDP
Catalog No. 5000295549
Encompass_Preferred
Punicalin is a natural polyphenolic compound isolated from Punica granatum (pomegranate) and used as a research reagent for antiviral, anti-inflammatory, and antioxidant studies. It is supplied at high reported purity with documented solubility and storage recommendations for both in vitro and in vivo applications.
  • High purity (≈99.9%).
  • Reported anti-HBV and broad antiviral activity.
  • Exhibits anti-inflammatory and antioxidant effects.
  • Reported to induce pyroptosis and inhibit carbonic anhydrase.
  • Documented solubility: soluble in DMSO and water; in vivo formulation guidance provided.
  • Packaged in small prepacked research quantities (example: 10 mg).
Catalog No. 50-002-95549 Supplier Medchemexpress LLC Supplier No. HYN063910MG
Punicalin is a natural polyphenolic compound isolated from Punica granatum (pomegranate) and used as a research reagent for antiviral, anti-inflammatory, and antioxidant studies. It is supplied at high reported purity with documented solubility and storage recommendations for both in vitro and in vivo applications.
  • High purity (≈99.9%).
  • Reported anti-HBV and broad antiviral activity.
  • Exhibits anti-inflammatory and antioxidant effects.
  • Reported to induce pyroptosis and inhibit carbonic anhydrase.
  • Documented solubility: soluble in DMSO and water; in vivo formulation guidance provided.
  • Packaged in small prepacked research quantities (example: 10 mg).