Learn More
Invivogen pUNO1 bearing the human STING gene, HAQ variant
SDP
Supplier:  Invivogen PUNO1HSTINGHAQ
Encompass
Human STING gene, R71H-G230A-R293Q variantSTING (stimulator of interferon genes; also known as TMEM173, MITA, MPYS, and ERIS) is essential for the IFN response to microbial or self-DNA, and acts as a direct sensor of cyclic dinucleotides (CDNs).CDNs are important messengers in bacteria, affecting numerous responses of the prokaryotic cell, but also in mammalian cells, acting as agonists of the innate immune response.
Catalog No. NC3450664