Learn More
ILEX LIFE SCIENCES LLC Biocolor Purple-Jelley Hyaluronan Assay, Large Economy Size Kit (400 assays)
SDP
Supplier:  ILEX LIFE SCIENCES LLC H2000
Encompass_Preferred
The Biocolor Purple-Jelley Hyaluronan Assay is a quantitative colorimetric dye-binding assay used to measure the quantity of hyaluronic acid (HA) in mammalian tissue samples. It is a dye-based method with a detection limit of 0.2 μg. The dye used is ‘Stains-all’. The assay manual includes methods for the user to remove tissue protein and sulfated glycosaminoglycans (sGAG) before measuring the isolated hyaluronic acid. Also included in the manual is an outline guide to the distribution of hyaluronic acid present in various animal tissues – heart, kidney, lung, leg muscle and skin. All values were obtained using this assay. In adult animals the hyaluronic acid percentage gradually increases during ageing as muscle and fat mass decrease. Sensitivity: 0.2 µg. Synonyms: hyaluronic acid, HA, Purple-Jelly Hyaluronan Assay
Catalog No. 50-212-0000