Learn More
Sigma Aldrich Fine Chemicals Biosciences Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 15 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT
Catalog No. NC2244952
Encompass
Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 15 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT
This item is not returnable. View return policy
Catalog No. NC2244952 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PURX120151KT
Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 15 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT