Learn More
Sigma Aldrich Fine Chemicals Biosciences Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 5 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT
Catalog No. NC1408834
Encompass
Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 5 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT
Catalog No. NC1408834 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PURX120051KT
Pur-A-Lyzer(TM) Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 5 Maxi dialysis tubes, MWCO 12-14 kDa | 1KT