Learn More
Sigma Aldrich Fine Chemicals Biosciences Pur-A-Lyzer(TM) Mega Dialysis Kit Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 20 mL displacement | 1KT
Catalog No. NC2216301
Encompass
Pur-A-Lyzer(TM) Mega Dialysis Kit Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 20 mL displacement | 1KT
This item is not returnable. View return policy
Catalog No. NC2216301 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PURG350201KT
Pur-A-Lyzer(TM) Mega Dialysis Kit Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 20 mL displacement | 1KT