Learn More
Sigma Aldrich Fine Chemicals Biosciences Pur A LyzerTM Maxi Dialysi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PURX121001KT
Encompass
The Pur-A-Lyzer Dialysis Kits are an innovative dialysis tubing that can be conveniently loaded and unloaded using any standard pipette tip while still allowing for 98% sample recovery yield. The revolutionary design of the Pur-A-Lyzer Tube makes it the only dialysis tool which enables true high throughput dialysis accommodating a large number of samples in a single buffer tank.The Pur-A-Lyzer Dialysis Tube allows rapid high performance extraction of macromolecules without contamination. It is ideal for purification of proteins automated protein sequencing peptide mapping and amino acid analysis.
Catalog No. 50-290-8939