Learn More
Sigma Aldrich Fine Chemicals Biosciences Pur-A-LyzerTM Mega Dialysi
Supplier: Sigma Aldrich Fine Chemicals Biosciences PURG100201KT
The Pur-A-Lyzer Dialysis Kits are an innovative dialysis tubing that can be conveniently loaded and unloaded using any standard pipette tip while still allowing for 98% sample recovery yield. The revolutionary design of the Pur-A-Lyzer Tube makes it the only dialysis tool which enables true high throughput dialysis accommodating a large number of samples in a single buffer tank.The Pur-A-Lyzer Dialysis Tube allows rapid high performance extraction of macromolecules without contamination. It is ideal for purification of proteins automated protein sequencing peptide mapping and amino acid analysis.