Learn More
Biolegend Purified anti-human GARP (LRRC32)
Supplier:  Biolegend 352502
Encompass_Preferred
Purified anti-human GARP (LRRC32) [7B11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. NC1723629