Learn More
Biolegend Purified anti-STAT1 Phospho (Tyr701)
Supplier:  Biolegend 666402
Encompass_Preferred
Purified anti-STAT1 Phospho (Tyr701) [A17012A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICFC, ICC; Size: 100 ug
Catalog No. NC1537121