Learn More
iQ Biosciences Purified Human CD3 Pan T Cells, 10M cells/vial
Catalog No. NC2216701
Encompass_Preferred
High-quality, high-viability (> 90%) human CD3 Pan T cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 99%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
This item is not returnable. View return policy
Catalog No. NC2216701 Supplier iQ Biosciences Supplier No. IQBHU1T10
High-quality, high-viability (> 90%) human CD3 Pan T cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 99%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.