Learn More
Grainger PUR Tread Al Core Wheel
Catalog No. 502766470
Encompass_Preferred
The Polyurethane Tread on Aluminum Stair Tread by Safe-T-Metal is a durable and slip-resistant stair tread designed for industrial and commercial use. It features an abrasive aluminum base with angled lugs for traction and a polyurethane top layer for comfort and noise reduction. This 16" x 48" tread can be installed on pan-filled stairs and landings to improve safety and meet code requirements. With its aluminum and polyurethane construction, it stands up well to heavy foot traffic while providing grip and reducing noise. The angled lugs allow water, dirt and debris to pass through.
Catalog No. 50-276-6470 Supplier Grainger Supplier No. 16V361
The Polyurethane Tread on Aluminum Stair Tread by Safe-T-Metal is a durable and slip-resistant stair tread designed for industrial and commercial use. It features an abrasive aluminum base with angled lugs for traction and a polyurethane top layer for comfort and noise reduction. This 16" x 48" tread can be installed on pan-filled stairs and landings to improve safety and meet code requirements. With its aluminum and polyurethane construction, it stands up well to heavy foot traffic while providing grip and reducing noise. The angled lugs allow water, dirt and debris to pass through.