Learn More
Grainger PUR Tread on Steel Core Wheel
Catalog No. 502767117
Encompass_Preferred
The Grainger 16V293 is a Dayton 1/3 HP 115/230V Dual Voltage Farm Duty Electric Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a 1075 RPM operating speed with a 56C ambient temperature rating. It features permanent split capacitor construction with ball bearings and a thermally protected design. The 16V293 measures approximately 9-1/2" in length by 5-3/4" in diameter and weighs 15 pounds. It can be used for a wide variety of farm equipment applications.
Catalog No. 50-276-7117 Supplier Grainger Supplier No. 16V293
The Grainger 16V293 is a Dayton 1/3 HP 115/230V Dual Voltage Farm Duty Electric Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a 1075 RPM operating speed with a 56C ambient temperature rating. It features permanent split capacitor construction with ball bearings and a thermally protected design. The 16V293 measures approximately 9-1/2" in length by 5-3/4" in diameter and weighs 15 pounds. It can be used for a wide variety of farm equipment applications.