Learn More
Julabo USA Inc PURA 10; 10L water bath, 1.2 kW heating power, +18 to +99.9 °C (with accessory lid 9970581; accessory cooling coil 9970534), 115V
SDP
Catalog No. 501904624
Encompass_Preferred
PURA water baths from JULABO. All PURA water baths have an integrated drain tap. Stainless steel bath tanks with a sloped bottom makes draining easy. Units include a false bottom yielding a flat surface for sample vessels. No heating element in the bath. Easy operation with bright LED. Operation in °F or °C with integrated timer. Working temperature range from +18°C to +99.9°C (with accessory cooling coil and bath tank lid). Models with filling volumes from 4.8 to 36L. Temperature stability of ± 0.15°C. Heating capacity up to 2 kW. Bright display. Integrated protection against running dry. Transparent lift-up bath covers for PURA Water Baths up to 100°C. The bath cover is made of Plexiglas; reduces water evaporation during incubation.
Catalog No. 50-190-4624 Supplier Julabo USA Inc Supplier No. 9550510.22
PURA water baths from JULABO. All PURA water baths have an integrated drain tap. Stainless steel bath tanks with a sloped bottom makes draining easy. Units include a false bottom yielding a flat surface for sample vessels. No heating element in the bath. Easy operation with bright LED. Operation in °F or °C with integrated timer. Working temperature range from +18°C to +99.9°C (with accessory cooling coil and bath tank lid). Models with filling volumes from 4.8 to 36L. Temperature stability of ± 0.15°C. Heating capacity up to 2 kW. Bright display. Integrated protection against running dry. Transparent lift-up bath covers for PURA Water Baths up to 100°C. The bath cover is made of Plexiglas; reduces water evaporation during incubation.