Learn More
Julabo USA Inc PURA 4; 4L water bath, 500 W heating power, RT to +99.9 °C (with accessory lid 9970580), 115V
SDP
Catalog No. 501904623
Encompass_Preferred
PURA water baths from JULABO. All PURA water baths have an integrated drain tap. Stainless steel bath tanks with a sloped bottom makes draining easy. Units include a false bottom yielding a flat surface for sample vessels. No heating element in the bath. Easy operation with bright LED. Operation in °F or °C with integrated timer. Working temperature range from +18°C to +99.9°C (with accessory cooling coil and bath tank lid). Models with filling volumes from 4.8 to 36L. Temperature stability of ± 0.15°C. Heating capacity up to 2 kW. Bright display. Integrated protection against running dry. Transparent lift-up bath covers for PURA Water Baths up to 100°C. The bath cover is made of Plexiglas; reduces water evaporation during incubation.
Catalog No. 50-190-4623 Supplier Julabo USA Inc Supplier No. 9550504.22
PURA water baths from JULABO. All PURA water baths have an integrated drain tap. Stainless steel bath tanks with a sloped bottom makes draining easy. Units include a false bottom yielding a flat surface for sample vessels. No heating element in the bath. Easy operation with bright LED. Operation in °F or °C with integrated timer. Working temperature range from +18°C to +99.9°C (with accessory cooling coil and bath tank lid). Models with filling volumes from 4.8 to 36L. Temperature stability of ± 0.15°C. Heating capacity up to 2 kW. Bright display. Integrated protection against running dry. Transparent lift-up bath covers for PURA Water Baths up to 100°C. The bath cover is made of Plexiglas; reduces water evaporation during incubation.