Learn More
Aviva Systems Biology Corp Pura Antibody - C-terminal region (ARP38689_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38689_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pura. It was validated on Western Blot. . Alternate names: CAGER-1, Pur-alpha, ssCRE-BPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-284-752