Learn More
Aviva Systems Biology Corp PURA Antibody - N-terminal region (ARP38688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PURA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PURA. Alternate names: PUR-ALPHA, PUR1, PURALPHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56755