Learn More
Aviva Systems Biology Corp PURA Antibody - N-terminal region (ARP33201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PURA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PURA. Alternate names: PUR1, MRD31, PURALPHA, PUR-ALPHAApplicationWB. Species reactivity: Human
Catalog No. 50-014-54916