Learn More
Moldex™ Pura-Fit™ Ecostation™ Ear Plug Refill
SDP
Catalog No. 19384374 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
33 dB
Catalog No. Noise Reduction Rating (NRR)
19-384-374 33 dB
1 options
Catalog No. 19-384-374 Supplier Moldex™ Supplier No. 6706
Add to Cart
Add to Cart
Convenient refill container for Pura-Fit™ Ecostation™ dispenser (6705). Pura-Fit™ ear plugs are tapered with a longer shape for easier insertion and grasping. Special foam is formulated for softness and comfort.
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape with longer length for easy insertion and removal
  • Non-irritating smooth surface
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility Yes
Packaging Type Refill
Pairs per Package 500 Pairs/Dispenser Refill
Plug Material Foam
Plug Color Green
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No