Learn More
Med Vet International McKesson Puracyn Plus Professional Antimicrobial Hydrogel, 3 oz., 186-6542, 6/Case
Supplier:  Med Vet International 1113215CS
Encompass_Preferred
Antimicrobial Hydrogel McKesson Puracyn Plus Professional 3 oz. NonSterile FDA-approved to add and maintain moisture in the wound bed and facilitate autolytic debridement of acute and chronic dermal lesions including itch and pain relief Intended for use by healthcare professionals to moisten the wound bed and facilitate autolytic debridement of acute and chronic dermal lesions Relieves itch and pain associated with dermal irritation, sores, injuries, and ulcers of dermal tissue, in addition to moistening and lubricating absorbent wound dressings Contains hypochlorous acid, a known antimicrobial, which serves as a preservative to inhibit the growth of microorganisms in the hydrogel Hydrogel maintains a moist wound environment, thereby encouraging autolytic debridement of the wound and relieving discomfort (i.e. itch and pain) associated with dermal irritation
Catalog No. 50-304-2924