Learn More
Med Vet International McKesson Puracyn Plus Professional Wound Irrigation Solution, 8.5oz, 186-6509, 6/Case
Catalog No. 503044439
Encompass_Preferred
Wound Cleanser McKesson Puracyn Plus Professional 8.5 oz. Pump Bottle NonSterile Antimicrobial For cleansing, irrigating, moistening, and debriding to remove wound debris from acute and chronic dermal lesions Non-cytotoxic, non-irritating and won't harm healthy tissue Convenient, ready-to-use formula requires no mixing or diluting provided in a pump spray bottle for easy, one-finger application Supports wound odor reduction Shelf life is up to 24 months Hypochlorous acid, a known antimicrobial, serves as a preservative to inhibit the growth of a broad range of microorganisms including staphylococcal species 8.5 oz. pump bottle
Catalog No. 50-304-4439 Supplier Med Vet International Supplier No. 1113211CS
Wound Cleanser McKesson Puracyn Plus Professional 8.5 oz. Pump Bottle NonSterile Antimicrobial For cleansing, irrigating, moistening, and debriding to remove wound debris from acute and chronic dermal lesions Non-cytotoxic, non-irritating and won't harm healthy tissue Convenient, ready-to-use formula requires no mixing or diluting provided in a pump spray bottle for easy, one-finger application Supports wound odor reduction Shelf life is up to 24 months Hypochlorous acid, a known antimicrobial, serves as a preservative to inhibit the growth of a broad range of microorganisms including staphylococcal species 8.5 oz. pump bottle