Learn More
Med Vet International McKesson Puracyn Plus Wound Irrigation Solution, 8½ oz. Pump Bottle, 186-6002, Each
Catalog No. 503044741
Encompass_Preferred
Wound Cleanser McKesson Puracyn Plus 8.5 oz. Pump Bottle NonSterile Antimicrobial Effectively cleans minor cuts, scrapes, sores and abrasions and prepares them to heal Alternative to commercial wound care products that are not biocompatible and may prolong the healing process Streamlines the process of caring for minor wounds and sores at home Advanced, super-oxygenated, pH-balanced hypochlorous solution to help set the stage for optimal healing conditions Non-cytotoxic, non-irritating, and won't harm healthy tissue Contains no antibiotics or steroids Contains no alcohol or iodine Ingredients: Hypochlorous acid (HOCI) (0.012%), sodium hypochlorite (NaOCI) (0.001%), electrolyzed water (H2O) (99.916%), sodium chloride (NaCI) (0.031%), phosphates (0.040%)
Catalog No. 50-304-4741 Supplier Med Vet International Supplier No. 1113216EA
Wound Cleanser McKesson Puracyn Plus 8.5 oz. Pump Bottle NonSterile Antimicrobial Effectively cleans minor cuts, scrapes, sores and abrasions and prepares them to heal Alternative to commercial wound care products that are not biocompatible and may prolong the healing process Streamlines the process of caring for minor wounds and sores at home Advanced, super-oxygenated, pH-balanced hypochlorous solution to help set the stage for optimal healing conditions Non-cytotoxic, non-irritating, and won't harm healthy tissue Contains no antibiotics or steroids Contains no alcohol or iodine Ingredients: Hypochlorous acid (HOCI) (0.012%), sodium hypochlorite (NaOCI) (0.001%), electrolyzed water (H2O) (99.916%), sodium chloride (NaCI) (0.031%), phosphates (0.040%)