Learn More
Med Vet International McKesson Puracyn Plus Professional Wound Irrigation Solution, 16.9oz, 186-6516, Each
Catalog No. 503044738
Encompass_Preferred
Wound Cleanser McKesson Puracyn Plus Professional 16.9 oz. Flip Top Bottle NonSterile Antimicrobial For cleansing, irrigating, moistening, and debriding to remove wound debris from acute and chronic dermal lesions Non-cytotoxic, non-irritating, and won't harm healthy tissue Convenient, ready-to-use formula requires no mixing or diluting and provided with a septum-capped bottle for easy application Supports wound odor reduction Shelf life of up to 24 months Featured hypochlorous acid, a known antimicrobial, serves as a preservative to inhibit the growth of a broad range of microorganisms including staphylococcal species 16.9 oz. flip top bottle
Catalog No. 50-304-4738 Supplier Med Vet International Supplier No. 1113212EA
Wound Cleanser McKesson Puracyn Plus Professional 16.9 oz. Flip Top Bottle NonSterile Antimicrobial For cleansing, irrigating, moistening, and debriding to remove wound debris from acute and chronic dermal lesions Non-cytotoxic, non-irritating, and won't harm healthy tissue Convenient, ready-to-use formula requires no mixing or diluting and provided with a septum-capped bottle for easy application Supports wound odor reduction Shelf life of up to 24 months Featured hypochlorous acid, a known antimicrobial, serves as a preservative to inhibit the growth of a broad range of microorganisms including staphylococcal species 16.9 oz. flip top bottle