Learn More
Air Science Usa Laminar Flow Cabinet 24, Vertical Airflow, ISO Class 5, 115V
SDP
Catalog No. NC2401257
Encompass_Preferred
The Purair Flow Laminar Flow Cabinet 24 is a compact vertical laminar flow hood that protects work surfaces and materials from contamination. It features ISO Class 5 air cleanliness and utilizes HEPA filtration, ensuring a sterile work zone. Unique attributes include an energy-efficient LED lighting system and a quiet, high-performance ebm-papst Ec centrifugal fan, making it ideal for laboratory settings with limited space
Catalog No. NC2401257 Supplier Air Science Usa Supplier No. FLOW24A
The Purair Flow Laminar Flow Cabinet 24 is a compact vertical laminar flow hood that protects work surfaces and materials from contamination. It features ISO Class 5 air cleanliness and utilizes HEPA filtration, ensuring a sterile work zone. Unique attributes include an energy-efficient LED lighting system and a quiet, high-performance ebm-papst Ec centrifugal fan, making it ideal for laboratory settings with limited space