Learn More
Air Science Usa PCR Workstation 48" Wide
SDP
Catalog No. NC1847408
Encompass_Preferred
The Purair PCR Workstation is a versatile laminar flow cabinet that ensures a contamination-free environment for PCR amplification and aseptic techniques. Its vertical laminar airflow, HEPA filtration, and built-in germicidal UV lamp provide maximum protection against particulate contamination. Constructed with durable materials and UV-absorbing panels for 360-degree visibility, it is ideal for labs needing reliable performance and accessibility, distinguishing it from conventional workstations
Catalog No. NC1847408 Supplier Air Science Usa Supplier No. PCR48A
The Purair PCR Workstation is a versatile laminar flow cabinet that ensures a contamination-free environment for PCR amplification and aseptic techniques. Its vertical laminar airflow, HEPA filtration, and built-in germicidal UV lamp provide maximum protection against particulate contamination. Constructed with durable materials and UV-absorbing panels for 360-degree visibility, it is ideal for labs needing reliable performance and accessibility, distinguishing it from conventional workstations