Learn More
Air Science Usa GP Plus Carbon Filter, for Solvents, Alcohols, Organics, General Odors, for all Ductless Fume Hoods
SDP
Catalog No. NC0107389
Encompass_Preferred
GP Plus! general purpose carbon filter for solvents, alcohols, organics, and general odors for all Purair Basic Ductless Fume Hoods. Models P524, P536 require qty 1, P548, require qty 2
Catalog No. NC0107389 Supplier Air Science Usa Supplier No. ASTS001
GP Plus! general purpose carbon filter for solvents, alcohols, organics, and general odors for all Purair Basic Ductless Fume Hoods. Models P524, P536 require qty 1, P548, require qty 2