Learn More
Aviva Systems Biology Corp PURB Antibody - N-terminal region (ARP41073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PURB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PURB. Alternate names: MGC126784, MGC126786, PURBETAApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57710