Learn More
Air Science Usa Air Science Purair Basic Ductless Fume Hood, Enhanced Safety, Advanced Filtration, 48 in
SDP
Supplier:  Air Science Usa P548XTA
Encompass_Preferred
The Air Science Purair Basic Ductless Fume Hood is designed for safe and efficient handling of hazardous vapors in lab and educational settings. Its ductless design eliminates the need for venting, allowing for flexible placement and portability. Equipped with advanced Multiplex™ Filtration Technology, it ensures enhanced safety by filtering harmful particles and fumes, making it an ideal choice for diverse laboratory applications.
Catalog No. NC3428165