Learn More
Biolegend Purified anti-human C3a/C3a(desArg)/C3
Supplier:  Biolegend 518101
Encompass_Preferred
Purified anti-human C3a/C3a(desArg)/C3 [K13/16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IP, WB; Size: 50 ug
Catalog No. 50-170-235