Learn More
Biolegend Purified anti-mouse CD43 Activation-Associated Glycoform
Supplier:  Biolegend 121202
Encompass_Preferred
Purified anti-mouse CD43 Activation-Associated Glycoform [1B11]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP, WB; Size: 200 ug
Catalog No. 50-163-902