Learn More
Biolegend Purified (azide-free) anti-α-Synuclein, 103-108
Supplier:  Biolegend 807801
Encompass_Preferred
Purified (azide-free) anti-α-Synuclein, 103-108 [4B12/Synuclein]; Isotype: Mouse IgG1; Reactivity: Human; Apps: IHC-P, ELISA, WB; Size: 200 uL
Catalog No. 50-102-9646