Learn More
Biolegend Purified anti-Acetylated Lysine
Supplier:  Biolegend 623401
Encompass_Preferred
Purified anti-Acetylated Lysine [15G10]; Isotype: Mouse IgG2b, κ; Reactivity: Acetylated lysine residues; Apps: WB, ICC; Size: 25 ug
Catalog No. 50-170-753